missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse SOST/Sclerostin Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 2 publications
POA
Specifications
Antigen | SOST/Sclerostin |
---|---|
Clone | 248121 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
SOST/Sclerostin Monoclonal specifically detects SOST/Sclerostin in Mouse samples. It is validated for Western Blot.Specifications
SOST/Sclerostin | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
AAI37834 | |
50964 | |
Mouse myeloma cell line NS0-derived recombinant mouse SOST Gln24-Tyr211 Accession # AAI37834 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse SOST in direct ELISAs and Western blots. In direct ELISAs, this antibody does not cross-react with recombinant human SOST. |
248121 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
sclerostin, VBCHsclerosteosis | |
SOST | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only