missing translation for 'onlineSavingsMsg'
Learn More

Sorbitol, BAKER ANALYZED™ Biochemical Reagent, J.T. Baker™

Catalog Number p-5391267
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
50-70-4
C6H14O6
182.172
FBPFZTCFMRRESA-JGWLITMVSA-N
d-sorbitol, sorbitol, d-glucitol, glucitol, l-gulitol, --sorbitol, diakarmon, glucarine, sorbol, d---sorbitol
5780
CHEBI:17924
(2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol
C(C(C(C(C(CO)O)O)O)O)O
This item is not returnable. View return policy
50-70-4
C6H14O6
182.172
FBPFZTCFMRRESA-JGWLITMVSA-N
d-sorbitol, sorbitol, d-glucitol, glucitol, l-gulitol, --sorbitol, diakarmon, glucarine, sorbol, d---sorbitol
5780
CHEBI:17924
(2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol
C(C(C(C(C(CO)O)O)O)O)O

High quality biochemicals for research use

Packaging Poly Bottle
Quantity 500 g
Specific Gravity 1L = 1.49kg
Formula Weight 182.17
Grade BAKER ANALYZED™ Biochemical Reagent
Chemical Name or Material Sorbitol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'