missing translation for 'onlineSavingsMsg'
Learn More

Sonepiprazole, Tocris Bioscience™

Selective D4 receptor antagonist

Brand:  Tocris (Bio-Techne) 418510

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13444304

  • £163.17 / 10mg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective, high affinity antagonist for the rat and human dopamine D4 receptor (Ki = 10 nM). Exhibits low affinity at other monoamine receptors (Ki > 2000 nM). Induces c-fos gene expression in medial prefrontal cortex in a similar manner to clozapine (Cat. No. 0444). Brain penetrant and orally available.
Specifications

Chemical Identifiers

170858-33-0
401.525
sonepiprazole, isochr-etpip-phso2nh2, sonepiprazole hydrate, chembl69759, 4-4-2-isochroman-1-yl ethyl piperazin-1-yl benzenesulfonamide, 4-4-2-1s-3,4-dihydro-1h-2-benzopyran-1-yl ethyl-1-piperazinyl-benzenesulfonamide, 4-4-2-1s-3,4-dihydro-1h-isochromen-1-yl ethyl piperazin-1-yl benzenesulfonamide, 4-4-2-1s-isochroman-1-yl ethyl piperazin-1-yl benzenesulfonamide hydrate, sonepiprazole inn, d01rae
4-[4-[2-[(1S)-3,4-dihydro-1H-isochromen-1-yl]ethyl]piperazin-1-yl]benzenesulfonamide
C21H27N3O3S
WNUQCGWXPNGORO-NRFANRHFSA-N
133079
C1COC(C2=CC=CC=C21)CCN3CCN(CC3)C4=CC=C(C=C4)S(=O)(=O)N

Specifications

10mg
170858-33-0
sonepiprazole, isochr-etpip-phso2nh2, sonepiprazole hydrate, chembl69759, 4-4-2-isochroman-1-yl ethyl piperazin-1-yl benzenesulfonamide, 4-4-2-1s-3,4-dihydro-1h-2-benzopyran-1-yl ethyl-1-piperazinyl-benzenesulfonamide, 4-4-2-1s-3,4-dihydro-1h-isochromen-1-yl ethyl piperazin-1-yl benzenesulfonamide, 4-4-2-1s-isochroman-1-yl ethyl piperazin-1-yl benzenesulfonamide hydrate, sonepiprazole inn, d01rae
C1COC(C2=CC=CC=C21)CCN3CCN(CC3)C4=CC=C(C=C4)S(=O)(=O)N
401.525
401.52
Sonepiprazole
C21H27N3O3S
WNUQCGWXPNGORO-NRFANRHFSA-N
4-[4-[2-[(1S)-3,4-dihydro-1H-isochromen-1-yl]ethyl]piperazin-1-yl]benzenesulfonamide
133079
>98%
Videos
Documents