Learn More
Somatostatin Monoclonal Antibody (ICDCLS), eFluor™ 660, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 50-9751-82
Code : NEW
Additional Details : Weight : 0.09500kg
Description
The hormone somatostatin has active 14 aa and 28 aa forms that are produced by alternate cleavage of the single preproprotein encoded by this gene. Somatostatin is expressed throughout the body and inhibits the release of numerous secondary hormones by binding to high-affinity G-protein-coupled somatostatin receptors. This hormone is an important regulator of the endocrine system through its interactions with pituitary growth hormone, thyroid stimulating hormone, and most hormones of the gastrointestinal tract. Somatostatin also affects rates of neurotransmission in the central nervous system and proliferation of both normal and tumorigenic cells.
Somatostatin Monoclonal antibody specifically detects Somatostatin in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
Somatostatin | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P60041, P60042, P61278 | |
SST | |
Affinity chromatography | |
RUO | |
20604, 24797, 6750 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
ICDCLS | |
eFluor 660 | |
SST | |
Antrin; Growth hormone release-inhibiting factor; Neuronostatin; NST; preprosomatostatin; prepro-somatostatin; SMST; SOM; Somatostatin; somatostatin 28; somatostatin-14; Somatostatin-28; somatotropin release-inhibiting factor; SRIF; SS; SS-14; SS-28; Sst; SST-14 | |
Mouse | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 κ |
For Research Use Only.