missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Solutions Aloe Sense Nitrile Gloves

Features a textured surface for dry and wet working conditions. Fisherbrand™ Solutions Aloe Sense Nitrile Gloves with aloe vera coating on the inner surface protect hands from drying, chapping and cracking.

£34.73 - £35.00

Specifications

Coating Material Aloe Vera
Color Blue, Green
Glove Cuff Style Beaded/Rolled
Hand Ambidextrous
Length (Metric) 240mm
View More Specs

Products 5
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
15103279 Fisherbrand™
Medium £35.00
Pack of 100 View alternative products
Save Up to 
»
In Stock 705
Add to basket
 
15193269 Fisherbrand™
Small £35.00
Pack of 100 View alternative products
Save Up to 
»
In Stock 245
Add to basket
 
15113279 Fisherbrand™
X-Large £35.00
Pack of 100 View alternative products
Save Up to 
»
In Stock 117
Add to basket
 
11772779 View accessory products Fisherbrand™
Large £35.00
Pack of 100 View alternative products
Save Up to 
»
In Stock 878
Add to basket
 
15123279 Fisherbrand™
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
Description

Description

These gloves feature a textured surface to provide ideal grip for dry and wet working conditions. Aloe vera coating on the inner surface protects hands from drying, chapping and cracking, making this glove ideal for prolonged use. Beaded cuff. Mint green for easy identification.
  • PPE Category III
  • Aloe vera coating protects from drying, chapping and cracking
  • Ideal for prolonged use
  • Textured surface provides good grip
  • Mint green
  • Beaded cuff
  • Disposable
  • Nitrile
Specifications

Specifications

Aloe Vera
Beaded/Rolled
240mm
Powder-free
20MPa
Blue, Green
Ambidextrous
Nitrile
Non-sterile
ISO 9001 QMS, FDA Biocompatibility Guidance, 510K Approved