missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Solid PTFE Stirring Rod

Chemically resistant, inert and leak-free

Brand:  Fisherbrand™

Code : 41

Additional Details : Weight : 0.06768kg

Product Code. 11782634

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

  • Chemically resistant, inert and leak-free
  • Use up to 280°C (540°F)
  • With steel core for maximum rigidity
Specifications

Specifications

Use up to 280°C (540°F)
8mm
300mm
PTFE (Polytetrafluoroethylene)
Videos
Documentation