missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Solid PTFE Stirring Rod

Can be used in temperatures up to 280°C (540°F). The PTFE stirring rod has a solid PTFE core. These rods are chemically resistant, inert, and leak-free.

Brand:  Fisherbrand™

Code : 41

Additional Details : Weight : 0.03000kg

Product Code. 11752634

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Chemically resistant, inert, and leak-free
  • Can be used in temperatures up to 280°C (540°F)
  • Solid PTFE rod with a tapered end
  • Will not scratch
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers