Learn More
Soldering flux, liquid, Thermo Scientific Chemicals
All purpose flux for general soldering
Brand: Thermo Scientific Alfa Aesar 045581.AP
Code : NEW
Additional Details : Weight : 0.00050kg Transport : UN number : 1840 Chem class : 8 Pack group : III
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Soldering flux | |
MFCD01868519 | |
500 mL |
1.26 g/mL | |
UN1840 | |
Liquid |
RUO – Research Use Only