missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Sodium zirconium oxide, 98%

Brand:  Alfa Aesar™ 018762.22

Code : NEW

Additional Details : CAS Number : 12201-48-8 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 11313477

  • £164.10 / 100g


Estimated Shipment date
from Supplier 25-04-2018
Add to basket

Specifications

Chemical Identifiers

Sodium zirconium oxide
185.2
Sodium zirconate
12201-48-8
MFCD00014245

Specifications

Sodium zirconium oxide
Powder
185.2
Sodium zirconate
White
12201-48-8
98%
MFCD00014245
−100 Mesh
100g
Videos
SDS