missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Tungstate Dihydrate, Honeywell™
Click to view available options
Quantity:
50 g
100 g
500 g
Packaging:
Poly bottle
Specifications
Melting Point | 698°C |
Color | Colorless |
pH | 9.0 to 11.5 (at 100.00g/L 20°C) |
Quantity | 500 g |
Linear Formula | Na2WO4 · 2H2O |
UN Number | NONH for all modes of transport |
Formula Weight | 329.85g/mol |
Odor | Odorless |
Packaging | Poly bottle |
Appearance | Crystalline |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'