missing translation for 'onlineSavingsMsg'
Learn More

Sodium trimethylsilanolate, 1M solution in dichloromethane, AcroSeal™, Thermo Scientific Chemicals

Catalog Number 10509778
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
18027-10-6, 75-09-2
C3H9NaOSi
112.18
MFCD00013926
HSNUIYJWTSJUMS-UHFFFAOYSA-N
sodium trimethylsilanolate, silanol, trimethyl-, sodium salt, trimethylsilanol sodium salt, silanol, 1,1,1-trimethyl-, sodium salt 1:1, silanol,1,1,1-trimethyl-, sodium salt 1:1, naotms, sodiooxytrimethylsilane, naosi me 3, trimethylsilyloxy sodium, sodium trimethylsilanolat
23672319
sodium;trimethyl(oxido)silane
C[Si](C)(C)[O-].[Na+]
This item is not returnable. View return policy
18027-10-6, 75-09-2
C3H9NaOSi
112.18
MFCD00013926
HSNUIYJWTSJUMS-UHFFFAOYSA-N
sodium trimethylsilanolate, silanol, trimethyl-, sodium salt, trimethylsilanol sodium salt, silanol, 1,1,1-trimethyl-, sodium salt 1:1, silanol,1,1,1-trimethyl-, sodium salt 1:1, naotms, sodiooxytrimethylsilane, naosi me 3, trimethylsilyloxy sodium, sodium trimethylsilanolat
23672319
sodium;trimethyl(oxido)silane
C[Si](C)(C)[O-].[Na+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 800 mL
Flash Point 63°C
Formula Weight 112.18
Packaging AcroSeal™ Glass Bottle
Specific Gravity 1.28
Density 1.2800g/mL
Chemical Name or Material Sodium trimethylsilanolate
compliance-icons compliance-icons
Product Identifier
  • Sodium trimethylsilanolate, 1M solution in dichloromethane
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H351-Suspected of causing cancer.
  • H336-May cause drowsiness or dizziness.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'