Learn More
Sodium triethylborohydride, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 200038000
Code : 29
Additional Details : CAS Number : 17979-81-6 Weight : 0.76200kg Transport : UN number : 3399 Chem class : 4.3 Pack group : I
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentSpecifications
Sodium triethylborohydride | |
17979-81-6,109-99-9 | |
0.8900g/mL | |
AcroSeal™ Glass bottle | |
NaB(C2H5)3H | |
800 mL | |
sodium triethylborohydride, sodium triethylborate, sodium triethylhydroborate, sodium triethylhydridoborate, sodium triethylhydroborate 1-, sodium triethylborohydride solution, sodium triethyl-? 2-boranuide, sodiumtriethylborohydride 1m in toluene | |
YDTZLEUIYNMRLQ-UHFFFAOYSA-N | |
sodium;triethylboron(1-) | |
23667700 | |
Solution |
1M solution in THF | |
Colorless | |
−17°C | |
C6H16BNa | |
MFCD00011704 | |
0.89 | |
Solubility in water: rzacts. | |
[Na+].CC[BH-](CC)CC | |
121.99 | |
121.99 |
RUO – Research Use Only