missing translation for 'onlineSavingsMsg'
Learn More

Sodium triethylborohydride, 1M solution in THF, AcroSeal™, ACROS Organics™

Brand:  Acros Organics 200031000

Code : 29

Additional Details : CAS Number : 17979-81-6 Weight : 0.13900kg Transport : UN number : 3399 Chem class : 1 Pack group : I

Quantity 100mL
Packaging AcroSeal™ Glass bottle

 View more versions of this product

Product Code. 10490471

  • £69.00 / 100mL


EU Stock 5
Add to basket

This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

Description

Description

Reduction reagent
Specifications

Chemical Identifiers

Sodium triethylborohydride
109-99-9
NaB(C2H5)3H
MFCD00011704
1M solution in THF
C6H16BNa
121.99

Specifications

Sodium triethylborohydride
109-99-9
Solution
NaB(C2H5)3H
MFCD00011704
Solubility in water: rzacts.
100mL
0.89
1M solution in THF
0.8900g/mL
C6H16BNa
121.99
AcroSeal™ Glass bottle
Colorless
−17°C
Videos
SDS