missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™

Brand:  Fisher Chemical J/7950/24

Code : 23

Additional Details : CAS Number : 10102-17-7 Weight : 10.00000kg

Packaging Bag in box
Quantity 10L

 View more versions of this product

Product Code. 10121910

Quantity Price
1 £82.24 / 10 L
10 £76.24 / 10 L
In stock, ready to ship: 23

Please to purchase this item. Need a web account? Register with us today!



Specifications

Chemical Identifiers

10102-17-7
248.172
PODWXQQNRWNDGD-UHFFFAOYSA-L
61475
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
H10Na2O8S2
149186
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
CHEBI:32150
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]

Specifications

10102-17-7
149186
PODWXQQNRWNDGD-UHFFFAOYSA-L
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
61475
Liquid
10L
H10Na2O8S2
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]
248.172
CHEBI:32150
Bag in box
Videos
SDS Documents