missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium thiosulfate pentahydrate, 99+%, for biochemistry

Catalog Number p-4813670
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy
10102-17-7
Na2O3S2·5H2O
248.18
PODWXQQNRWNDGD-UHFFFAOYSA-L
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
61475
CHEBI:32150
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]
This item is not returnable. View return policy
10102-17-7
Na2O3S2·5H2O
248.18
PODWXQQNRWNDGD-UHFFFAOYSA-L
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
61475
CHEBI:32150
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 48.5°C
Quantity 250 g
Assay Percent Range 99.0 to 101% (Iodimetry)
Linear Formula Na2S2O3·5H2O
Merck Index 15,8821
Solubility Information (1 M in water) Clear colorless
Formula Weight 248.18
Percent Purity ≥99%
Grade Biochemical
Packaging Plastic bottle
Specific Gravity 1.72
Density 1.7200g/mL
Chemical Name or Material Sodium thiosulfate pentahydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'