missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiosulfate pentahydrate, 99.5+%, ACS reagent, Thermo Scientific Chemicals

Catalog Number p-3735163
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle
10102-17-7
MFCD00149186
PODWXQQNRWNDGD-UHFFFAOYSA-L
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
61475
CHEBI:32150
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]
10102-17-7
MFCD00149186
PODWXQQNRWNDGD-UHFFFAOYSA-L
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
61475
CHEBI:32150
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY
Color White
pH 6.0 to 8.4 (5% soln. at 25°C)
Physical Form Crystals
Quantity 1 kg
Assay Percent Range 99.5+%
Molecular Formula H10Na2O8S2
Linear Formula Na2S2O3·5H2O
Merck Index 15, 8821
Molecular Weight (g/mol) 248.172
Formula Weight 248.18
Percent Purity 99.5 to 101.0%
Grade ACS Reagent
Packaging Plastic bottle
Decomposition Information >45°C
Density 1.7200g/mL
Chemical Name or Material Sodium thiosulfate pentahydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'