missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiosulfate, Acculute Standard Volumetric Solution, Final Concentration 0.1N, Thermo Scientific Chemicals

Catalog Number p-7080475
Click to view available options
Quantity:
1 U
6 U
This item is not returnable. View return policy
7772-98-7
Na2O3S2
158.10
MFCD00003499
AKHNMLFCWUSKQB-UHFFFAOYSA-L
sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt
24477
[Na+].[Na+].[O-]S([S-])(=O)=O
This item is not returnable. View return policy
7772-98-7
Na2O3S2
158.10
MFCD00003499
AKHNMLFCWUSKQB-UHFFFAOYSA-L
sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt
24477
[Na+].[Na+].[O-]S([S-])(=O)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Linear Formula Na2S2O3
IUPAC Name disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane
Formula Weight 158.1
Quantity 1 U
Physical Form Liquid
Chemical Name or Material Sodium thiosulfate Acculute Standard Volumetric Solution, Final Concentration 0.1N

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'