missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate, 5-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog Number p-5390961
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
10102-17-7
H10Na2O8S2
248.172
PODWXQQNRWNDGD-UHFFFAOYSA-L
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
61475
CHEBI:32150
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]
This item is not returnable. View return policy
10102-17-7
H10Na2O8S2
248.172
PODWXQQNRWNDGD-UHFFFAOYSA-L
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
61475
CHEBI:32150
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]

High quality chemicals for laboratory and specialized industrial use

Physical Form Crystals
Quantity 500 g
Formula Weight 248.18
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 1.75kg
Chemical Name or Material Sodium Thiosulfate, 5-Hydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'