missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium thiosulfate, 0.01N Standardized Solution, Thermo Scientific Chemicals
77.11 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Use promo code "21741" to get your promotional price.
Click to view available options
Quantity:
1 L
4 L
20 L
Description
Sodium thiosulfate, 0.01N standardized solution is used in volumetric analysis to estimate the concentration of certain compounds in solution (hydrogen peroxide) as well as chlorine content in bleaching powder and water. It is used as a titrant in iodometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Linear Formula | Na2S2O3 |
Concentration | 0.01N |
Solubility Information | Immiscible with alcohol. |
Formula Weight | 158.1 |
Quantity | 1 L |
Physical Form | Liquid |
Chemical Name or Material | Sodium thiosulfate, Standardized Solution |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'