missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiocyanate, 98+%, for analysis, Thermo Scientific Chemicals

Catalog Number 13458608
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy
540-72-7
CNNaS
81.07
MFCD00011123
VGTPCRGMBIAPIM-UHFFFAOYSA-M
sodium thiocyanate, sodium rhodanate, sodium sulfocyanate, sodium rhodanide, thiocyanic acid, sodium salt, thiocyanate sodium, scyan, sodium isothiocyanate, nascn, haimased
516871
CHEBI:30952
sodium;thiocyanate
[Na+].[S-]C#N
This item is not returnable. View return policy
540-72-7
CNNaS
81.07
MFCD00011123
VGTPCRGMBIAPIM-UHFFFAOYSA-M
sodium thiocyanate, sodium rhodanate, sodium sulfocyanate, sodium rhodanide, thiocyanic acid, sodium salt, thiocyanate sodium, scyan, sodium isothiocyanate, nascn, haimased
516871
CHEBI:30952
sodium;thiocyanate
[Na+].[S-]C#N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 287.0°C
Quantity 250 g
Assay Percent Range 98% min. (Argentometry)
Linear Formula NaSCN
Merck Index 15,948
Solubility Information Solubility in water: 1390g/L (21°C). Other solubilities: soluble in alcohol, freely soluble in acetone
Formula Weight 81.07
Percent Purity ≥98%
Grade Analytical
Packaging Glass bottle
Chemical Name or Material Sodium thiocyanate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'