Learn More
Sodium tetraphenylborate, ACS, 99.5% min, Thermo Scientific Chemicals
Use promo code "21615" to get your promotional price.
Description
Sodium tetraphenylborate is used as a precipitating reagent for potassium, as a protecting agent to protect amino acids. It is used in the preparation of N-acylammonium salts. It finds application as a phenyl donor in palladium-catalyzed cross-coupling reactions to prepare arylalkenes and biaryl compounds. It is used in inorganic and organometallic chemistry due to its solubility in nonpolar solvents. Furthermore, it is involved in the isolation of complexes having dinitrogen ligands.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | >300°C |
Color | White |
pH | 8 |
Linear Formula | NaB(C6H5)4 |
Quantity | 2 g |
UN Number | UN2811 |
Beilstein | 3599783 |
Sensitivity | Light sensitive |
Merck Index | 14,8690 |
Solubility Information | Soluble in water and ethanol. |
Show More |
RUO – Research Use Only