missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium tetrakis(4-fluorophenyl)borate dihydrate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
Sodium tetrakis(4-fluorophenyl)borate dihydrate, is used as a reagent for the two-phase titration of nonionic surfactants in the presence of anionic surfactants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium tetrakis(4-fluorophenyl)borate dihydrate, is used as a reagent for the two-phase titration of nonionic surfactants in the presence of anionic surfactants.
Notes
Hygroscopic. Incompatible withstrong oxidizing agents.
Sodium tetrakis(4-fluorophenyl)borate dihydrate, is used as a reagent for the two-phase titration of nonionic surfactants in the presence of anionic surfactants.
Notes
Hygroscopic. Incompatible withstrong oxidizing agents.
Specifications
Quantity | 250 mg |
Beilstein | 4285808 |
Formula Weight | 450.21 (414.18 Anhydrous) |
Percent Purity | 98% |
Chemical Name or Material | Sodium tetrakis(4-fluorophenyl)borate dihydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'