missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Sodium tetrakis(1-imidazolyl)borate, 97%

Brand:  Alfa Aesar™ H55081.14

Code : NEW

Additional Details : CAS Number : 68146-65-6 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15401228

  • £401.85 / 25g


Estimated Shipment date
from Supplier 09-03-2018
Add to basket

Specifications

Chemical Identifiers

Sodium tetrakis(1-imidazolyl)borate
68146-65-6
302.08
MFCD00012083

Specifications

Sodium tetrakis(1-imidazolyl)borate
68146-65-6
97%
MFCD00012083
25g
Solid
302.08
Videos
SDS