missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium tetrachloropalladate(II) hydrate, Premion™, 99.999% (metals basis), Pd 30% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Sodium tetrachloropalladate is used in organ metallic chemistry for the preparation of phosphine complexes of palladium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Red to Brown |
Physical Form | Crystalline |
Quantity | 1 g |
Linear Formula | Na2PdCl4·xH2O (x≈3) |
Solubility Information | Soluble in water and ethyl alcohol. |
Formula Weight | 294.20 (Anhydrous) |
Percent Purity | 99.999% |
Grade | Premion™ |
Concentration or Composition (by Analyte or Components) | Pd 30% min |
Sensitivity | Hygroscopic |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'