missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium tetrachloropalladate(II), approx. 36% Pd, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Brown to Orange or Brown-Green |
Physical Form | Powder and Granules, Crystals |
Quantity | 25 g |
Assay Percent Range | 36% |
Linear Formula | Na2PdCl4 |
Fieser | 05,626; 06,45; 09,207; 16,309; 01,778; 04,365; 05,498; 06,443; 07,274; 15,13; 16,356 |
Solubility Information | Solubility in water: soluble |
IUPAC Name | disodium tetrachloropalladiumbis(ylium) |
Formula Weight | 294.21 |
Percent Purity | ∼36% Pd |
Show More |
Safety and Handling
Product Identifier
- Sodium tetrachloropalladate(II)
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Substances and mixtures corrosive to metals Category 1
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
- H290-May be corrosive to metals.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'