missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetrachloroaurate(III) dihydrate, Premion™, 99.99% (metals basis), Au 49-50%, Thermo Scientific Chemicals

Catalog Number p-4412865
Click to view available options
Quantity:
1 g
5 g
13874-02-7
AuCl4H4NaO2
397.79
MFCD00149162
OEERDTKOVZFOHF-UHFFFAOYSA-J
sodium tetrachloroaurate iii dihydrate, sodium;tetrachlorogold 1-;dihydrate, sodiumtetrachloroaurate iii dihydrate, sodium dihydrate tetrachloroaurate
18949434
O.O.[Na+].Cl[Au-](Cl)(Cl)Cl
13874-02-7
AuCl4H4NaO2
397.79
MFCD00149162
OEERDTKOVZFOHF-UHFFFAOYSA-J
sodium tetrachloroaurate iii dihydrate, sodium;tetrachlorogold 1-;dihydrate, sodiumtetrachloroaurate iii dihydrate, sodium dihydrate tetrachloroaurate
18949434
O.O.[Na+].Cl[Au-](Cl)(Cl)Cl

It is used in chemo-therapeutics. It is employed as a catalyst in the removal of tert-butyl(dimethyl)silyl (TBS) protecting groups. It is selective in the presence of aromatic TBS ethers, aliphatic triisopropylsilyl ethers, aliphatic tert-butyl(diphenyl)silyl ethers, or sterically hindered aliphatic TBS ethers. With excess catalyst, TBS ethers can also be transformed into 4-methoxybenzyl ethers or methyl ethers in one pot.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 100°C (decomposition)
Physical Form Crystalline
Quantity 1 g
Assay Percent Range 99.99% (metals basis)
Solubility Information Soluble in water,alcohol and ether.
IUPAC Name sodium tetrachlorogolduide dihydrate
Formula Weight 397.80 (361.77 Anhydrous)
Density 0.8 g/mL
Chemical Name or Material Sodium tetrachloroaurate(III) dihydrate
compliance-icons compliance-icons
Product Identifier
  • Sodium tetrachloroaurate(III) dihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Aurate(1-), tetrachloro-, sodium, dihydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'