missing translation for 'onlineSavingsMsg'
Learn More

Sodium Tetraborate Decahydrate, Reagent Grade, ≥99.5%, Honeywell™

Catalog Number 15637710
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Drum with inliner
Poly Bottle
This item is not returnable. View return policy
1303-96-4
MFCD00149193
This item is not returnable. View return policy
1303-96-4
MFCD00149193

Reagent Grade,≥99.5%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Boiling Point 1575°C
Melting Point 62°C
Color Colorless
pH 9 to 10 (at 20°C)
Physical Form Solid
Quantity 500 g
Linear Formula Na2B4O7 · 10H2O
UN Number NONH for all modes of transport
Formula Weight 381.37g/mol
Percent Purity ≥99.5%
Odor Odorless
Packaging Poly Bottle
Density 1.73g/mL at 25°C
Chemical Name or Material Sodium tetraborate decahydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'