missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium tetraborate decahydrate, 99.5+%, ACS reagent, Thermo Scientific Chemicals
16.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
12.93 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "22914" to get your promotional price.
12.93 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 75.0°C |
Color | White |
pH | 9.15 to 9.20 (0.01M soln. at 25°C) |
Physical Form | Crystalline Powder |
Quantity | 25 g |
Assay Percent Range | 99.5+% |
Linear Formula | Na2B4O7·10H2O |
Fieser | 04,461 |
Merck Index | 15, 8726 |
Solubility Information | Solubility in water: 60g/L (20°C). Other solubilities: soluble in glycerol, insoluble in alcohol |
Show More |
Safety and Handling
Product Identifier
- Sodium tetraborate decahydrate
- Danger
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
- H319-Causes serious eye irritation.
- H360FD-May damage fertility. May damage the unborn child.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'