missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetraborate, 98%, for analysis, anhydrous, Thermo Scientific Chemicals

Catalog Number 11904331
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1330-43-4
B4Na2O7
201.21
MFCD00081185
NXSVMNYRVBQCMO-UHFFFAOYSA-N
Borax, fused
disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
[Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1330-43-4
B4Na2O7
201.21
MFCD00081185
NXSVMNYRVBQCMO-UHFFFAOYSA-N
Borax, fused
disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
[Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1575.0°C
Melting Point 741.0°C
Color Orange
Physical Form Crystalline Powder and/or Crystals
Quantity 2.5 kg
Assay Percent Range 98%
Linear Formula Na2B4O7
Fieser 04,461; 15,295
Merck Index 15,8726
Solubility Information Solubility in water: 26g/L (20°C). Other solubilities: insoluble in alcohol
Formula Weight 201.22
Percent Purity 98%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Sodium tetraborate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'