missing translation for 'onlineSavingsMsg'
Learn More

Sodium telluride, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7051687
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
12034-41-2
Na2Te
173.58
MFCD00053400
MQRWPMGRGIILKQ-UHFFFAOYSA-N
sodium telluride, disodium telluride, sodium telluridosodium, hydrotelluric acid, sodium salt
82837
sodiotellanylsodium
[Na][Te][Na]
This item is not returnable. View return policy
12034-41-2
Na2Te
173.58
MFCD00053400
MQRWPMGRGIILKQ-UHFFFAOYSA-N
sodium telluride, disodium telluride, sodium telluridosodium, hydrotelluric acid, sodium salt
82837
sodiotellanylsodium
[Na][Te][Na]

 

Sodium telluride is used as a semiconductor, as a reducing agent and to prepare organic tellurium compounds. It is used in the reduction of aromatic carbonyl compounds to alcohols. It is also used in the preparation of pyrrolo[2,3-d]pyrimidines (7-deaza-9H-purines) from aromatic nitriles. It is used to prepare nanocrystalline zinc telluride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY
Melting Point 953°C
Color White
Physical Form Powder
Quantity 2 g
UN Number UN3134
Solubility Information Highly soluble in water.
Formula Weight 173.58
Percent Purity 99.9%
Odor Odorless
Sensitivity Moisture sensitive
Density 2.90 g/mL
Assay (metals basis)
Chemical Name or Material Sodium telluride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'