missing translation for 'onlineSavingsMsg'
Learn More

Sodium Tartrate, Dihydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

£176.63

Chemical Identifiers

CAS 6106-24-7
Molecular Formula C4H10Na2O8
Molecular Weight (g/mol) 232.096
InChI Key YAPIZISBZUXUIH-DGFHWNFOSA-N
PubChem CID 131855972
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid;sodium;dihydrate
SMILES C(C(C(=O)O)O)(C(=O)O)O.O.O.[Na].[Na]
View More Specs

Products 1
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15598184
View Documents
JT Baker
3931-01
500g Poly Bottle
£176.63
500g
View alternative products
Save Up to 
»
Estimated Shipment
06-01-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

    Suitable for Standardization of Karl Fischer Reagents
Specifications

Chemical Identifiers

6106-24-7
232.096
131855972
C(C(C(=O)O)O)(C(=O)O)O.O.O.[Na].[Na]
C4H10Na2O8
YAPIZISBZUXUIH-DGFHWNFOSA-N
(2R,3R)-2,3-dihydroxybutanedioic acid;sodium;dihydrate

Specifications

Sodium Tartrate
500g
6106-24-7
C4H10Na2O8
C(C(C(=O)O)O)(C(=O)O)O.O.O.[Na].[Na]
232.096
230.08
BAKER ANALYZED™ ACS Reagent
1L = 1.82kg
Dihydrate
Poly Bottle
YAPIZISBZUXUIH-DGFHWNFOSA-N
(2R,3R)-2,3-dihydroxybutanedioic acid;sodium;dihydrate
131855972
Crystals
SDS