Learn More
Sodium Tartrate Dihydrate (Colorless Crystals), Fisher BioReagents
Brand: Fisher Bioreagents BP352-500
Code : 32
Additional Details : CAS Number : 6106-24-7 Weight : 0.55000kg
Description
Glass Bottle; 500g
This sodium salt is used in buffers for molecular biology and cell culture applications.Specifications
Sodium Tartrate Dihydrate | |
White | |
15.61 to 15.71% (at 150°C) | |
C4H8Na2O8 | |
500 g | |
unii-dia7c37aow, disodium tartrate dihydrate, disodium l-+-tartrate dihydrate, dia7c37aow, disodium +-tartrate dihydrate, sodium l-+-tartrate dihydrate, butanedioic acid, 2,3-dihydroxy-2r,3r-, disodium salt, dihydrate, disodium tartarate dihydrate, sodium tartrate nf, sodium tartrate dihydrate mi | |
FGJLAJMGHXGFDE-UDMGOBQRNA-L | |
(2R,3R)-2,3-dihydroxybutanedioic acid;sodium;dihydrate | |
131855972 | |
99.0 to 101.0% |
6106-24-7,868-18-8 | |
7 to 9 | |
Glass Bottle | |
MFCD00150035 | |
15, 8812 | |
≤0.005% Insoluble Matter | |
O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O | |
230.08 | |
230.08 | |
Powder or Solid |