missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium sulfite, Honeywell Fluka™
Click to view available options
Quantity:
250 g
2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 2.5 kg |
Linear Formula | Na2SO3 |
UN Number | NONH for all modes of transport |
Formula Weight | 126.04g/mol |
Percent Purity | ≥98% |
Chemical Name or Material | Sodium sulfite |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'