missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Sodium sulfide, tech., 90+%, H2O 5% max

Brand:  Alfa Aesar™ 088697.03

Code : NEW

Additional Details : CAS Number : 12034-39-8 Weight : 0.00100kg Transport : UN number : 1385 Chem class : 1b Pack group : II

Quantity 1g

 View more versions of this product

Product Code. 11329339

  • £14.00 / 1g


Estimated Shipment date
from Supplier 13-02-2018
Add to basket

Description

Description

  • For reducing organic nitro compounds, in the manufacture of sulfur dyes, in ore flotation, and in the preparation of metal sulfide finishes
Specifications

Chemical Identifiers

Sodium sulfide
Technical Grade
174.24
UN1385
12034-39-8
Na2S4
MFCD00014249

Specifications

Sodium sulfide
Technical Grade
≥90%
174.24
UN1385
H2O 5% max
Moisture Sensitive & Hygroscopic
275°C
12034-39-8
Powder
Na2S4
MFCD00014249
Soluble in water and alcohol
Like rotten eggs
Pale brown
1g
Videos
SDS