missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfide hydrate, tech., Thermo Scientific Chemicals

Catalog Number p-7048855
Click to view available options
Quantity:
500 g
2 kg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
27610-45-3
Na2S
78.04
MFCD00149183
GRVFOGOEDUUMBP-UHFFFAOYSA-N
45051681
[Na+].[Na+].[S--]
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
27610-45-3
Na2S
78.04
MFCD00149183
GRVFOGOEDUUMBP-UHFFFAOYSA-N
45051681
[Na+].[Na+].[S--]

Sodium sulfide hydrate is used as an oxygen scavenger in water treatment, bleaching agent in textile industry, desulfurising and as a dechlorinating agent and in leather industry, sulfonation and sulfomethylation agent in chemical industry. It is involved in the production of rubber chemicals and sulfur dyes. It is also used to prepare 3-nitroaniline from 1,3-dinitrobenzene and thioethers from alkyl halide. It plays an important role in other applications such as ore flotation, oil recovery, making dyes, and detergent. In addition, it is used in pulp and paper industry in the Kraft process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∽50°C (decomposition)
Physical Form Flakes
Quantity 500 g
UN Number UN1849
Merck Index 14,8681
Solubility Information Soluble in water; Slightly soluble in alcohol; Insoluble in etherSoluble in water; Slightly soluble in alcohol; Insoluble in ether.
IUPAC Name disodium sulfanediide
Formula Weight 78.04 (anhydrous)
Grade Technical
Sensitivity Hygroscopic; Light sensitive
Density 1.427 g/mL
Chemical Name or Material Sodium sulfide hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'