missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfate, Puriss., meets analytical spec. of BP, Ph. Eur., USP, anhydrous, 99.0-100.5% (calc. to the dried substance), Honeywell Fluka™
BIOPHARMA

Catalog Number p-7071812
14.18 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.


Click to view available options
Quantity:
1 kg
2.5 kg
25 kg
50 kg
6 x 1 kg
6 x 2.5 kg
100 kg
7757-82-6
Na2O4S
142.04
MFCD00003504
PMZURENOXWZQFD-UHFFFAOYSA-L
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
24436
CHEBI:32149
[Na+].[Na+].[O-]S([O-])(=O)=O
7757-82-6
Na2O4S
142.04
MFCD00003504
PMZURENOXWZQFD-UHFFFAOYSA-L
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
24436
CHEBI:32149
[Na+].[Na+].[O-]S([O-])(=O)=O

Puriss., meets analytical specification of BP, Ph. Eur., USP, anhydrous, 99.0-100.5% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Physical Form Anhydrous
Quantity 1 kg
Linear Formula Na2SO4
UN Number NONH for all modes of transport
IUPAC Name disodium;sulfate
Formula Weight 142.04g/mol
Percent Purity 99 to 100.5% (calc. to the dried substance)
Grade puriss.
Density 2.68g/mL at 25°C
Chemical Name or Material Sodium Sulfate, Meets analytical specification of Ph. Eur.Bp, USP
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'