missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfate Anhydrous, 99+%, Extra Pure, Fisher Chemical™

Catalog Number 10142780
83.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
500 g
3 kg
5 kg
7757-82-6
Na2O4S
142.04
MFCD00003504
PMZURENOXWZQFD-UHFFFAOYSA-L
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
24436
CHEBI:32149
[Na+].[Na+].[O-]S([O-])(=O)=O
7757-82-6
Na2O4S
142.04
MFCD00003504
PMZURENOXWZQFD-UHFFFAOYSA-L
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
24436
CHEBI:32149
[Na+].[Na+].[O-]S([O-])(=O)=O

Sodium Sulfate Anhydrous, 99+%, Extra Pure

TRUSTED_SUSTAINABILITY
Melting Point 884°C
Color White
pH 5 to 8
Physical Form Solid
Quantity 3 kg
IUPAC Name disodium sulfate
Formula Weight 142.04g/mol
Packaging Plastic powder jar
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'