missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfate, 99+%, for analysis, powder, anhydrous, Thermo Scientific Chemicals

Catalog Number p-222005
18.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
7757-82-6
Na2O4S
142.04
MFCD00003504
PMZURENOXWZQFD-UHFFFAOYSA-L
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
24436
CHEBI:32149
[Na+].[Na+].[O-]S([O-])(=O)=O
7757-82-6
Na2O4S
142.04
MFCD00003504
PMZURENOXWZQFD-UHFFFAOYSA-L
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
24436
CHEBI:32149
[Na+].[Na+].[O-]S([O-])(=O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point >1700.0°C
Melting Point 884.0°C
Color White
pH 5 to 8 (5% soln. at 20°C)
Physical Form Fine Crystalline Powder
Quantity 500 g
Assay Percent Range 99+%
Linear Formula Na2SO4
Fieser 01,1103
Merck Index 15, 8809
Solubility Information Solubility in water: 185g/L. Other solubilities: soluble in glycerol and hydrogen iodide, insoluble in alcohol
IUPAC Name disodium;sulfate
Formula Weight 142.04
Percent Purity ≥99%
Grade Analytical
Loss on Ignition 0.5% max. (800°C)
Packaging Plastic bottle
Decomposition Information Decomposition Information: >890°C
Density 2.68
Chemical Name or Material Sodium sulfate, 99%, p.a.
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'