missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfanilate hydrate, 97%, Thermo Scientific Chemicals

Catalog Number 10396731
Click to view available options
250g; Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS 123333-70-0
Melting Point 288°C
Color Brown or Pink to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (on dry substance)
Packaging Glass bottle
Molecular Formula C6H6NNaO3S
MDL Number MFCD00151043
Quantity 250 g
Synonym sodium sulfanilate hydrate, sodium 4-aminobenzenesulfonate hydrate, sulfanilic acid sodium salt hydrate, 4-aminobenzenesulfonic acid, sodium salt hydrate, sodium 4-aminobenzenesulfonate hydrate 1:1:x, acmc-209aoo, c6h6nso3.na.h2o, sodium4-aminobenzenesulfonatehydrate, p-anilinesulfonic acid sodium
Solubility Information Solubility in water: >20g/L
InChI Key KSVSZLXDULFGDQ-UHFFFAOYSA-M
SMILES [Na+].NC1=CC=C(C=C1)S([O-])(=O)=O
IUPAC Name sodium;4-aminobenzenesulfonate;hydrate
Molecular Weight (g/mol) 195.17
PubChem CID 23672401
Formula Weight 195.17
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Sodium sulfanilate hydrate, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'