Learn More
Sodium-silica gel, 35-40% alkali metal in silica gel, Stage II, Thermo Scientific™
Reductively Produces Radical Anions and Dianions, Dechlorinates Alkyl and Aryl Halides and Desulfurize Compounds. Also Ideal For Reduction of Water To Produce Hydrogen Gas, Solvent Drying and Anti-Oxidant
Brand: Thermo Scientific Alfa Aesar 044894.06
Code : NEW
Additional Details : Weight : 0.00500kg Transport : UN number : 2813 Chem class : 4.3 Pack group : III
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Sodium-silica gel | |
Black | |
5 g | |
Moisture sensitive |
35-40% alkali metal in silica gel, Stage II | |
Powder | |
MFCD01941718 | |
0.7 |