Learn More
Sodium-silica gel, 35-40% alkali metal in silica gel, Stage I, Thermo Scientific™
Reductively Produces Radical Anions and Dianions, Dechlorinates Alkyl and Aryl Halides and Desulfurize Compounds
£47.02 - £562.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15466625
|
Thermo Scientific Alfa Aesar
044893.14 |
25 g |
£204.00
25g |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15456625
|
Thermo Scientific Alfa Aesar
044893.06 |
5 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15476625
|
Thermo Scientific Alfa Aesar
044893.22 |
100 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||