missing translation for 'onlineSavingsMsg'
Learn More

Sodium selenite, anhydrous, 99% min, typically 99.75% min (metals basis), Thermo Scientific Chemicals

Catalog Number p-7049046
Click to view available options
Quantity:
100 g
500 g
10102-18-8
Na2O3Se
172.948
MFCD00003489
BVTBRVFYZUCAKH-UHFFFAOYSA-L
sodium selenite, disodium selenite, natriumselenit, selenious acid, disodium salt, natriumselenit german, disodium selenium trioxide, unii-hiw548rq3w, selenious acid disodium salt, ccris 1260, hsdb 768
24934
CHEBI:48843
disodium;selenite
[O-][Se](=O)[O-].[Na+].[Na+]
10102-18-8
Na2O3Se
172.948
MFCD00003489
BVTBRVFYZUCAKH-UHFFFAOYSA-L
sodium selenite, disodium selenite, natriumselenit, selenious acid, disodium salt, natriumselenit german, disodium selenium trioxide, unii-hiw548rq3w, selenious acid disodium salt, ccris 1260, hsdb 768
24934
CHEBI:48843
disodium;selenite
[O-][Se](=O)[O-].[Na+].[Na+]

In glass manufacturing

In glass manufacturingSodium selenite is used in the manufacture of colorless glass. It acts as an ingredient in some food supplements. It is an animal feed used in pet foods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >350°C
Color White
pH 9
Physical Form Powder
Quantity 100 g
Linear Formula Na2SeO3
UN Number UN2630
Merck Index 14,8673
Solubility Information Freely soluble in water; Insoluble in alcoholSoluble in water; Insoluble in alcohol and ethanol.
Formula Weight 172.94
Percent Purity ≤99%
Odor Odorless
Sensitivity Hygroscopic
Assay Typically ≤99.75% (metals basis)
Chemical Name or Material Sodium selenite, Anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'