missing translation for 'onlineSavingsMsg'
Learn More

Sodium selenate decahydrate, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number 11316479
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
10102-23-5
H20Na2O14Se
369.10
MFCD00149174
TYVNFQRMDPFYPC-UHFFFAOYSA-L
sodium selenate decahydrate, unii-0cjn4029eb, disodium decahydrate selenate, disodium selenate decahydrate, acmc-1c452, sodium selenate decahydrate, technical grade, selenic acid, disodiumsalt, decahydrate 9ci, sodium selenate decahydrate trace metals basis
202320
disodium;selenate;decahydrate
O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-][Se]([O-])(=O)=O
This item is not returnable. View return policy
10102-23-5
H20Na2O14Se
369.10
MFCD00149174
TYVNFQRMDPFYPC-UHFFFAOYSA-L
sodium selenate decahydrate, unii-0cjn4029eb, disodium decahydrate selenate, disodium selenate decahydrate, acmc-1c452, sodium selenate decahydrate, technical grade, selenic acid, disodiumsalt, decahydrate 9ci, sodium selenate decahydrate trace metals basis
202320
disodium;selenate;decahydrate
O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-][Se]([O-])(=O)=O

Sodium selenate decahydrate is used as a feed additive, as a mineral supplement, as analytical reagents and in horticultural applications. Further, it is used in manufacturing of glass to counter the color caused by iron impurities. It is also employed in the prevention of abnormal tissue growth. In addition to this, it is used as a micronutrient which prevents a deficiency of copper, manganese and selenium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 35°C (decomposition)
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 99.9% (metals basis)
UN Number UN2630
Merck Index 14,8671
Solubility Information Soluble in water.
Formula Weight 369.09 (188.94 Anhydrous)
Percent Purity 99.9%
Density 1.6 g/mL
Assay (metals basis)
Chemical Name or Material Sodium selenate decahydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'