missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Pyruvate, BAKER ANALYZED™ Reagent, J.T. Baker™
Click to view available options
Quantity:
1 kg
Packaging:
Wide Mouth Amber Glass Bottle
Specifications
Quantity | 1 kg |
Formula Weight | 110.05 |
Grade | BAKER ANALYZED™ Reagent |
Packaging | Wide Mouth Amber Glass Bottle |
Chemical Name or Material | Sodium Pyruvate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'