missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Sodium potassium (K2Na)-silica gel, 35-40% alkali metal in silica gel, Stage I

Brand:  Alfa Aesar™ 044890.22

Code : NEW

Additional Details : Weight : 0.10000kg Transport : UN number : 2813 Chem class : 4.3 Pack group : II

Quantity 100g

 View more versions of this product

Product Code. 15463915

  • £502.80 / 100g


Estimated Shipment date
from Supplier 31-05-2018
Add to basket

Description

Description

  • Reductively produce radical anions and dianions, dechlorinate alkyl and aryl halides and desulfurize compounds
Specifications

Chemical Identifiers

Sodium potassium (K2Na)-silica gel, Stage I
MFCD08705348
UN2813

Specifications

Sodium potassium (K2Na)-silica gel, Stage I
Powder
UN2813
35 to 40% alkali metal in silica gel
Moisture Sensitive
0.7g/mL
MFCD08705348
Black
100g
Videos
SDS