missing translation for 'onlineSavingsMsg'
Learn More

Sodium, plasma standard solution, Specpure™ Na 1000μg/mL, Thermo Scientific Chemicals

Catalog Number p-4904545
24.86 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
50 mL
100 mL
500 mL
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
Na2CO3 in 5% HNO3
MFCD00085307
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
Na2CO3 in 5% HNO3
MFCD00085307

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 50 mL
Vapor Pressure 23hPa (17mm Hg) at 20°C
Physical Form Liquid
Chemical Name or Material Na 1000μg/mL

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'