missing translation for 'onlineSavingsMsg'
Learn More

Sodium, plasma standard solution, Specpure™ Na 10,000μg/mL, Thermo Scientific Chemicals

Catalog Number 11312328
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Na2CO3 in 5% HNO3
MFCD00085307
Na2CO3 in 5% HNO3
MFCD00085307

Sodium, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 50 mL
Concentration or Composition (by Analyte or Components) Nitric acid: 5%; Sodium carbonate: 2.3%; Water: 92.7%
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Solubility Miscible with water
Physical Form Liquid
Chemical Name or Material Sodium, plasma standard solution

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'