missing translation for 'onlineSavingsMsg'
Learn More

Sodium pivalate hydrate, 99%, Thermo Scientific Chemicals

Catalog Number 10541483
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
143174-36-1
C5H9NaO2
124.12
MFCD00150782
SJRDNQOIQZOVQD-UHFFFAOYSA-M
sodium trimethylacetate hydrate, sodium pivalate hydrate, sodium hydrate pivalate, propanoic acid, 2,2-dimethyl-, sodium salt, hydrate, acmc-20ajn9, sodiumtrimethylacetatehydrate, c5h9o2.na.h2o, trimethylacetic acid sodium salt hydrate, sodium 2,2-dimethylpropanoate-water 1/1/1
23666172
sodium;2,2-dimethylpropanoate;hydrate
[Na+].CC(C)(C)C([O-])=O
This item is not returnable. View return policy
143174-36-1
C5H9NaO2
124.12
MFCD00150782
SJRDNQOIQZOVQD-UHFFFAOYSA-M
sodium trimethylacetate hydrate, sodium pivalate hydrate, sodium hydrate pivalate, propanoic acid, 2,2-dimethyl-, sodium salt, hydrate, acmc-20ajn9, sodiumtrimethylacetatehydrate, c5h9o2.na.h2o, trimethylacetic acid sodium salt hydrate, sodium 2,2-dimethylpropanoate-water 1/1/1
23666172
sodium;2,2-dimethylpropanoate;hydrate
[Na+].CC(C)(C)C([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula (CH3)3CCO2Na·xH2O
Quantity 10 g
Formula Weight 142.13
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material Sodium pivalate hydrate
compliance-icons
Product Identifier
  • Sodium pivalate hydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'