missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphonoformate hexahydrate, 98+%, Thermo Scientific Chemicals

Catalog Number p-7027776
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
34156-56-4
CH12Na3O11P
300.04
MFCD00150176
ILRVASBWNRYBFD-UHFFFAOYSA-K
foscarnet sodium hexahydrate, sodium phosphonatoformate hexahydrate, unii-964ys0oog1, foscarnet trisodium hexahydrate, phosphonoformic acid trisodium salt hexahydrate, trisodium phosphonoformte hexahydrate, sodium phosphonoformate tribasic hexahydrate, phosphonoformic acid, trisodium salt, hexahydrate, trisodium foscarnet 3-hexahydrate, formic acid, phosphono-, trisodium salt, hexahydrate
169569
CHEBI:60269
O.O.O.O.O.O.[Na+].[Na+].[Na+].[O-]C(=O)P([O-])([O-])=O
This item is not returnable. View return policy
34156-56-4
CH12Na3O11P
300.04
MFCD00150176
ILRVASBWNRYBFD-UHFFFAOYSA-K
foscarnet sodium hexahydrate, sodium phosphonatoformate hexahydrate, unii-964ys0oog1, foscarnet trisodium hexahydrate, phosphonoformic acid trisodium salt hexahydrate, trisodium phosphonoformte hexahydrate, sodium phosphonoformate tribasic hexahydrate, phosphonoformic acid, trisodium salt, hexahydrate, trisodium foscarnet 3-hexahydrate, formic acid, phosphono-, trisodium salt, hexahydrate
169569
CHEBI:60269
O.O.O.O.O.O.[Na+].[Na+].[Na+].[O-]C(=O)P([O-])([O-])=O

Sodium phosphonoformate hexahydrate acts as an antiviral agent and reverse transcriptase inhibitor. It is also used to inhibit viral DNA polymerase. Further, it is used as a type II Pi transporter inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium phosphonoformate hexahydrate acts as an antiviral agent and reverse transcriptase inhibitor. It is also used to inhibit viral DNA polymerase. Further, it is used as a type II Pi transporter inhibitor.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Quantity 1 g
Solubility Information Soluble in water.
IUPAC Name trisodium;phosphonatoformate;hexahydrate
Formula Weight 300.08 (191.98 Anhydrous)
Percent Purity ≥98%
Chemical Name or Material Sodium phosphonoformate hexahydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'