missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Sodium phosphate, tribasic dodecahydrate, 98%, for biochemistry
Click to view available options
Quantity:
500 g
2.5 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
Specifications
Melting Point | 75.0°C |
Quantity | 500 g |
Assay Percent Range | 98% |
Linear Formula | Na3PO4·12H2O |
Merck Index | 15,8792 |
Solubility Information | Solubility in water: 240g/L (20°C). Other solubilities: practically insoluble in alcohol |
Formula Weight | 380.13 |
Percent Purity | 98% |
Grade | Biochemical |
Packaging | Plastic bottle |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'